missing translation for 'onlineSavingsMsg'
Learn More

2,3-Pentanedione 97.0+%, TCI America™

Catalog No. P005125ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P005125ML 25 mL
P0051100ML 100 mL
P0051500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P005125ML Supplier TCI America Supplier No. P005125ML

Chemical Identifiers

CAS 600-14-6
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00009313
InChI Key TZMFJUDUGYTVRY-UHFFFAOYSA-N
Synonym 2,3-pentanedione, acetylpropionyl, acetyl propionyl, 2,3-pentadione, acetylpropionyl van, unii-k4wbe45scm, acetyl propionyl natural, 2,3-pentane-dione, pentan-2,3-dione, ethyl methyl diketone
PubChem CID 11747
ChEBI CHEBI:52774
IUPAC Name pentane-2,3-dione
SMILES CCC(=O)C(C)=O

Specifications

Melting Point -52°C
Color Yellow
Boiling Point 108°C
Quantity 25 mL
UN Number 1224
Formula Weight 100.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Pentanedione
EINECSNumber (2)-0563&(2)-2474
RTECSNumber SA1850000
TSCA Yes
Recommended Storage Freezer