missing translation for 'onlineSavingsMsg'
Learn More

2,3-Pentanedione, 97%

Catalog No. AC221470250
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC221470250 25 mL Glass bottle
AC221471000 100 mL Glass bottle
AC221475000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC221470250 Supplier Thermo Scientific Chemicals Supplier No. 221470250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 600-14-6 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 600-14-6
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00009313
InChI Key TZMFJUDUGYTVRY-UHFFFAOYSA-N
Synonym 2,3-pentanedione, acetylpropionyl, acetyl propionyl, 2,3-pentadione, acetylpropionyl van, unii-k4wbe45scm, acetyl propionyl natural, 2,3-pentane-dione, pentan-2,3-dione, ethyl methyl diketone
PubChem CID 11747
ChEBI CHEBI:52774
IUPAC Name pentane-2,3-dione
SMILES CCC(=O)C(C)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -52.0°C
Color Yellow
Density 0.9580g/mL
Boiling Point 110.0°C to 112.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4020 to 1.4060
Linear Formula C2H5COCOCH3
Quantity 25 mL
Beilstein 01,776
Specific Gravity 0.958
Solubility Information Solubility in water: 60g/L (15°C). Other solubilities: miscible with ethanol, glycerol and, propylene glycol, ether
Formula Weight 100.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 3-Pentanedione, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-984-8
RTECSNumber SA1850000
TSCA TSCA

RUO – Research Use Only