missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2',3'-O-Isopropylideneadenosine, 98%

Catalog No. AC122720050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC122720050 5 g Glass bottle
AC122720250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC122720050 Supplier Thermo Scientific Chemicals Supplier No. 122720050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 362-75-4 | C13H17N5O4 | 307.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 362-75-4
Molecular Formula C13H17N5O4
Molecular Weight (g/mol) 307.31
MDL Number MFCD00005756
InChI Key LCCLUOXEZAHUNS-AUWRGFAENA-N
Synonym 2',3'-o-isopropylideneadenosine, 3ar,4r,6r,6ar-6-6-amino-9h-purin-9-yl-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxol-4-yl methanol, 2',3'-isopropylideneadenosine, 3ar,4r,6r,6ar-6-6-amino-9h-purin-9-yl-2,2-dimethyl-tetrahydro-2h-furo 3,4-d 1,3 dioxol-4-yl methanol, 2 ,3-o-isopropylideneadenosine, 1r,2r,4r,5r-4-6-aminopurin-9-yl-7,7-dimethyl-3,6,8-trioxabicyclo 3.3.0 oc t-2-yl methan-1-ol, 3ar,4r,6r,6ar-4-6-aminopurin-9-yl-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro 3,4-d 1,3 dioxol-6-yl methanol, 3ar,4r,6r,6ar-6-6-aminopurin-9-yl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl methanol, pubchem14185
PubChem CID 2723654
IUPAC Name [(3aR,4R,6R,6aR)-4-(6-aminopurin-9-yl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-6-yl]methanol
SMILES CC1(C)O[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O1)N1C=NC2=C(N)N=CN=C12

Specifications

Absorbance 0.75 to 0.81 (A250/A260),0.13 to 0.17 (A280/A260),0.01 (A290/A260) max
CAS Min % 97.5
CAS Max % 100.0
Melting Point 221.0°C to 222.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Spectrophotometry 96% min. (lambda max. 259nm,pH 7) molar absorptivity 15400+/-4%,on dry substance
Formula Weight 307.3
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2', 3'-O-Isopropylideneadenosine, 98%
Show More Show Less
EINECSNumber 206-650-3
TSCA TSCA

RUO – Research Use Only