accessibility menu, dialog, popup

UserName

CAS RN 362-75-4

IUPAC Name: [(3aR,4R,6R,6aR)-6-(6-amino-9H-purin-9-yl)-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-yl]methanol
Synonyms: [(3aR,4R,6R,6aR)-6-(6-aminopurin-9-yl)-2,2-dimethyl-tetrahydrofuro[3,4-d][1,3]dioxol-4-yl]methanol
Molecular Weight (g/mol): 307.31
Molecular Formula: C13H17N5O4
InChi Key: LCCLUOXEZAHUNS-AUWRGFAENA-N
SMILES: CC1(C)O[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O1)N1C=NC2=C(N)N=CN=C12
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (2)
  • (4)
  • (3)
  • (3)
  • (4)