missing translation for 'onlineSavingsMsg'
Learn More

(-)-2,3-O-Isopropylidene-D-threitol 98.0+%, TCI America™

Catalog No. I03755G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03755G 5 g
I03751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I03755G Supplier TCI America Supplier No. I03755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 73346-74-4
Molecular Formula C7H14O4
Molecular Weight (g/mol) 162.185
MDL Number MFCD00009761
InChI Key INVRLGIKFANLFP-PHDIDXHHSA-N
Synonym --2,3-o-isopropylidene-d-threitol, 2,3-o-isopropylidene-d-threitol, 4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-diyl dimethanol, 2-3-o-isopropylidene-d-threitol, 4r,5r---2,2-dimethyl-1,3-dioxolane-4,5-dimethanol, trans-4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-diyl dimethanol, 4r,5r-5-hydroxymethyl-2,2-dimethyl-1,3-dioxolan-4-yl methanol, 4r,5r-4,5-bis hydroxymethyl-2,2-dimethyl-1,3-dioxolane, pubchem6768
PubChem CID 785244
IUPAC Name [(4R,5R)-5-(hydroxymethyl)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol
SMILES CC1(OC(C(O1)CO)CO)C

Specifications

Melting Point 50°C
Color White
Boiling Point 96°C
Quantity 5 g
Formula Weight 162.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (-)-2,3-O-Isopropylidene-D-threitol
TSCA No
Recommended Storage Freezer