CAS RN 73346-74-4
IUPAC Name:
[(4R,5R)-5-(hydroxymethyl)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol
Synonyms:
[(4R,5R)-5-(hydroxymethyl)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol
Molecular Weight (g/mol):
162.19
Molecular Formula:
C7H14O4
InChi Key:
INVRLGIKFANLFP-PHDIDXHHSA-N
SMILES:
CC1(C)O[C@H](CO)[C@@H](CO)O1
1
–
1
of
1
results
1
(-)-2,3-O-Isopropylidene-D-threitol 98.0+%, TCI America™
CAS: 73346-74-4 Molecular Formula: C7H14O4 Molecular Weight (g/mol): 162.185 MDL Number: MFCD00009761 InChI Key: INVRLGIKFANLFP-PHDIDXHHSA-N Synonym: --2,3-o-isopropylidene-d-threitol,2,3-o-isopropylidene-d-threitol,4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-diyl dimethanol,2-3-o-isopropylidene-d-threitol,4r,5r---2,2-dimethyl-1,3-dioxolane-4,5-dimethanol,trans-4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-diyl dimethanol,4r,5r-5-hydroxymethyl-2,2-dimethyl-1,3-dioxolan-4-yl methanol,4r,5r-4,5-bis hydroxymethyl-2,2-dimethyl-1,3-dioxolane,pubchem6768 PubChem CID: 785244 IUPAC Name: [(4R,5R)-5-(hydroxymethyl)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol SMILES: CC1(OC(C(O1)CO)CO)C