Learn More
2,3-O-Isopropylidene-D-ribonic acid-1,4-lactone, 97+%
CAS: 30725-00-9 | C8H12O5 | 188.18 g/mol
Supplier: Thermo Scientific Chemicals J66442ME
Description
2,3-O-Isopropylidene-D-ribonic acid-1,4-lactone is used for the synthesis of C-nucleosides, prostanoids and GABA analogs via homochiral 2-piperidones. It is the starting material for the preparation of biochemical compounds including C-nucleosides, such as neplanocin A, and GABA analogs, via homochiral 2-piperidones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,3-O-Isopropylidene-D-ribonic acid-1,4-lactone is used for the synthesis of C-nucleosides, prostanoids and GABA analogs via homochiral 2-piperidones. It is the starting material for the preparation of biochemical compounds including C-nucleosides, such as neplanocin A, and GABA analogs, via homochiral 2-piperidones.
Solubility
Soluble in methanol at 50mg/ml, acetone, dichloromethane, ethyl Acetate.
Notes
Soluble in methanol at 50mg/ml
Specifications
| 2,3-O-Isopropylidene-D-ribonic acid-1,4-lactone | |
| 135°C to 138°C | |
| C8H12O5 | |
| 500 mg | |
| 3ar,6r,6ar-6-hydroxymethyl-2,2-dimethyldihydrofuro 3,4-d 1,3 dioxol-4 3ah-one, 2,3-o-isopropylidene-d-ribonic gamma-lactone, 2,3-o-isopropylidene-d-ribonic gamma-lactone, 2,3-o-isopropylidene-d-ribono-1,4-lactone, 3ar,6r,6ar-6-hydroxymethyl-2,2-dimethyl-dihydro-3ah-furo 3,4-d 1,3 dioxol-4-one, d-2,3-isopropylidene lyxono-1,4-lactone, d-2,3-isopropylidene lyxono-1,4-lactone, nhhkfjcwlppncn-hsuxutppsa, 2,3-o-isopropylidene-d-ribonic, 2,3-o-isopropylidene-d-ribonic, a-lactone | |
| NHHKFJCWLPPNCN-UHFFFAOYNA-N | |
| CC1(C)OC2C(CO)OC(=O)C2O1 | |
| 188.18 | |
| 188.18 | |
| Powder |
| 30725-00-9 | |
| White | |
| MFCD00080793 | |
| 10090 | |
| Soluble in methanol at 50mg/ml,acetone,dichloromethane,ethyl Acetate. | |
| −68° (c=2 in pyridine) | |
| 6-(hydroxymethyl)-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-one | |
| 1268067 | |
| 98% |
Chemical Identifiers
| 30725-00-9 | |
| 188.18 | |
| NHHKFJCWLPPNCN-UHFFFAOYNA-N | |
| 1268067 | |
| CC1(C)OC2C(CO)OC(=O)C2O1 |
| C8H12O5 | |
| MFCD00080793 | |
| 3ar,6r,6ar-6-hydroxymethyl-2,2-dimethyldihydrofuro 3,4-d 1,3 dioxol-4 3ah-one, 2,3-o-isopropylidene-d-ribonic gamma-lactone, 2,3-o-isopropylidene-d-ribonic gamma-lactone, 2,3-o-isopropylidene-d-ribono-1,4-lactone, 3ar,6r,6ar-6-hydroxymethyl-2,2-dimethyl-dihydro-3ah-furo 3,4-d 1,3 dioxol-4-one, d-2,3-isopropylidene lyxono-1,4-lactone, d-2,3-isopropylidene lyxono-1,4-lactone, nhhkfjcwlppncn-hsuxutppsa, 2,3-o-isopropylidene-d-ribonic, 2,3-o-isopropylidene-d-ribonic, a-lactone | |
| 6-(hydroxymethyl)-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-one |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only