missing translation for 'onlineSavingsMsg'
Learn More

2,3-O-Isopropylidene-D-erythronolactone 98.0+%, TCI America™

Catalog No. I04541G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I04541G Supplier TCI America Supplier No. I04541G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25581-41-3
Molecular Formula C7H10O4
Molecular Weight (g/mol) 158.15
MDL Number MFCD00134440
InChI Key WHPSMBYLYRPVGU-UHFFFAOYNA-N
Synonym 2,3-o-isopropylidene-d-erythronolactone, --2,3-o-isopropylidene-d-erythronolactone, 3ar,6ar-2,2-dimethyldihydrofuro 3,4-d 1,3 dioxol-4 3ah-one, 3ar,6ar-2,2-dimethyl-6,6a-dihydro-3ah-furo 3,4-d 1,3 dioxol-4-one, 3ar,6ar-2,2-dimethyl-dihydro-3ah-furo 3,4-d 1,3 dioxol-4-one, pubchem19725, 2,3-o-isopropylidene-d-erythronolactol, d-2,3-o-isopropylidene-erythronic acid, a-lactone, 2-o,3-o-isopropylidene-d-erythronic acid gamma-lactone
PubChem CID 386843
IUPAC Name 2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-one
SMILES CC1(C)OC2COC(=O)C2O1

Specifications

Melting Point 68°C
Quantity 1 g
Formula Weight 158.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-O-Isopropylidene-D-erythronolactone