accessibility menu, dialog, popup

UserName

CAS RN 25581-41-3

IUPAC Name: 2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-one
Synonyms: 2,2-dimethyl-dihydro-3aH-furo[3,4-d][1,3]dioxol-4-one
Molecular Weight (g/mol): 158.15
Molecular Formula: C7H10O4
InChi Key: WHPSMBYLYRPVGU-UHFFFAOYNA-N
SMILES: CC1(C)OC2COC(=O)C2O1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)