missing translation for 'onlineSavingsMsg'
Learn More

2,3-Norbornanedicarboxylic Acid 98.0+%, TCI America™

Catalog No. N07535G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07535G 5 g
N075325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N07535G Supplier TCI America Supplier No. N07535G

Chemical Identifiers

CAS 1724-08-9
Molecular Formula C9H12O4
Molecular Weight (g/mol) 184.19
MDL Number MFCD00520577
InChI Key IVVOCRBADNIWDM-UHFFFAOYNA-N
Synonym Bicyclo[2.2.1]heptane-2,3-dicarboxylic Acid
PubChem CID 99835
IUPAC Name bicyclo[2.2.1]heptane-2,3-dicarboxylic acid
SMILES OC(=O)C1C2CCC(C2)C1C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 184.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Norbornanedicarboxylic Acid
RTECSNumber RB7168000
TSCA No