accessibility menu, dialog, popup

UserName

HOOHOO

CAS RN 1724-08-9

IUPAC Name: bicyclo[2.2.1]heptane-2,3-dicarboxylic acid
Synonyms: bicyclo[2.2.1]heptane-2,3-dicarboxylic acid
Molecular Weight (g/mol): 184.19
Molecular Formula: C9H12O4
InChi Key: IVVOCRBADNIWDM-UHFFFAOYNA-N
SMILES: OC(=O)C1C2CCC(C2)C1C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results