missing translation for 'onlineSavingsMsg'
Learn More

2,3-Naphthalenedicarboxaldehyde, 95%

Catalog No. p-4323221
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC294721000 100 mg Glass Bottle
AC294725000 500 mg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC294721000 Supplier Thermo Scientific Chemicals Supplier No. 294721000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7149-49-7 | C12H8O2 | 184.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7149-49-7
Molecular Formula C12H8O2
Molecular Weight (g/mol) 184.19
InChI Key ZIPLKLQPLOWLTM-UHFFFAOYSA-N
Synonym 2,3-naphthalenedicarboxaldehyde, 2,3-naphthalenedialdehyde, naphthalene-2,3-dicarboxaldehyde, 2,3-naphthalenedicarbaldehyde, naphthalene-2,3-dialdehyde, 2,3-nda, 2,3-diformylnaphthalene, 2,3-naphthalene dicarboxaldehyde, nnaphthalene-2,3-dicarboxaldehyde, naphthalene-2,3-dicarboxaldehyde nda
PubChem CID 96400
IUPAC Name naphthalene-2,3-dicarbaldehyde
SMILES C1=CC=C2C=C(C(=CC2=C1)C=O)C=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 134.0°C to 135.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Linear Formula C10H6(CHO)2
Quantity 100 mg
Formula Weight 184.19
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3-Naphthalenedicarboxaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only