missing translation for 'onlineSavingsMsg'
Learn More

2,3-(Methylenedioxy)benzaldehyde 97.0+%, TCI America™

$123.03 - $398.90

Chemical Identifiers

CAS 7797-83-3
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.133
MDL Number MFCD00016902
InChI Key QZMQKPGVXNSITP-UHFFFAOYSA-N
Synonym benzo d 1,3 dioxole-4-carbaldehyde, 4-formyl-1,3-benzodioxole, 2,3-methylenedioxy benzaldehyde, 1,3-benzodioxole-4-carboxaldehyde, 2h-1,3-benzodioxole-4-carbaldehyde, benzo-1,3-dioxole-4-carboxaldehyde, 2,3-methylenedioxybenzaldehyde, benzo 1,3 dioxole-4-carbaldehyde, 2h-benzo d 1,3-dioxolene-4-carbaldehyde, acmc-209pcy
PubChem CID 82264
IUPAC Name 1,3-benzodioxole-4-carbaldehyde
SMILES C1OC2=CC=CC(=C2O1)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
M13781G
View Documents
TCI America
M13781G
1 g
Each for $123.03
Only null left
 
M13785G
View Documents
TCI America
M13785G
5 g
Each for $398.90
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7797-83-3
150.133
QZMQKPGVXNSITP-UHFFFAOYSA-N
82264
C1OC2=CC=CC(=C2O1)C=O
C8H6O3
MFCD00016902
benzo d 1,3 dioxole-4-carbaldehyde, 4-formyl-1,3-benzodioxole, 2,3-methylenedioxy benzaldehyde, 1,3-benzodioxole-4-carboxaldehyde, 2h-1,3-benzodioxole-4-carbaldehyde, benzo-1,3-dioxole-4-carboxaldehyde, 2,3-methylenedioxybenzaldehyde, benzo 1,3 dioxole-4-carbaldehyde, 2h-benzo d 1,3-dioxolene-4-carbaldehyde, acmc-209pcy
1,3-benzodioxole-4-carbaldehyde

Specifications

7797-83-3
C8H6O3
1 g
QZMQKPGVXNSITP-UHFFFAOYSA-N
1,3-benzodioxole-4-carbaldehyde
82264
≥97.0% (GC)
2,3-(Methylenedioxy)benzaldehyde
Yellow
MFCD00016902
benzo d 1,3 dioxole-4-carbaldehyde, 4-formyl-1,3-benzodioxole, 2,3-methylenedioxy benzaldehyde, 1,3-benzodioxole-4-carboxaldehyde, 2h-1,3-benzodioxole-4-carbaldehyde, benzo-1,3-dioxole-4-carboxaldehyde, 2,3-methylenedioxybenzaldehyde, benzo 1,3 dioxole-4-carbaldehyde, 2h-benzo d 1,3-dioxolene-4-carbaldehyde, acmc-209pcy
C1OC2=CC=CC(=C2O1)C=O
150.133
150.13
Crystalline Lumps
Videos
Safety and Handling

Safety and Handling

RTECSNumber : DF4912450

TSCA : No

SDS
Documents

Documents