accessibility menu, dialog, popup

UserName

OOO

CAS RN 7797-83-3

IUPAC Name: 2H-1,3-benzodioxole-4-carbaldehyde
Synonyms: 2H-1,3-benzodioxole-4-carbaldehyde
Molecular Weight (g/mol): 150.13
Molecular Formula: C8H6O3
InChi Key: QZMQKPGVXNSITP-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC2=C1OCO2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results