missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-(Methylenedioxy)benzaldehyde 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 150.13 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 2,3-(Methylenedioxy)benzaldehyde |
Safety and Handling
| RTECSNumber | DF4912450 |
| TSCA | No |