missing translation for 'onlineSavingsMsg'
Learn More

2,3-(Methylenedioxy)benzaldehyde 97.0+%, TCI America™

Catalog No. M13781G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13781G 1 g
M13785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13781G Supplier TCI America Supplier No. M13781G
Only null left

Chemical Identifiers

CAS 7797-83-3
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.133
MDL Number MFCD00016902
InChI Key QZMQKPGVXNSITP-UHFFFAOYSA-N
Synonym benzo d 1,3 dioxole-4-carbaldehyde, 4-formyl-1,3-benzodioxole, 2,3-methylenedioxy benzaldehyde, 1,3-benzodioxole-4-carboxaldehyde, 2h-1,3-benzodioxole-4-carbaldehyde, benzo-1,3-dioxole-4-carboxaldehyde, 2,3-methylenedioxybenzaldehyde, benzo 1,3 dioxole-4-carbaldehyde, 2h-benzo d 1,3-dioxolene-4-carbaldehyde, acmc-209pcy
PubChem CID 82264
IUPAC Name 1,3-benzodioxole-4-carbaldehyde
SMILES C1OC2=CC=CC(=C2O1)C=O

Specifications

Color Yellow
Quantity 1 g
Formula Weight 150.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,3-(Methylenedioxy)benzaldehyde
RTECSNumber DF4912450
TSCA No