missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Methoxyphenyl)ethylamine 98.0+%, TCI America™

Catalog No. m18915g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
M18915G 5 g
M189125G 25 g
2 options

Catalog No. M18915G

Supplier: TCI America M18915G

Only null left

Chemical Identifiers

CAS 2039-67-0
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD00008187
InChI Key WJBMRZAHTUFBGE-UHFFFAOYSA-N
Synonym 3-methoxyphenethylamine, 3-methoxyphenylethylamine, 2-3-methoxyphenyl ethanamine, 2-3-methoxyphenyl ethylamine, m-methoxyphenethylamine, m-methoxyphenylethylamine, 2-3-methoxyphenyl ethan-1-amine, benzeneethanamine, 3-methoxy, phenethylamine, m-methoxy, 2-3-methoxy-phenyl-ethylamine
PubChem CID 74866
IUPAC Name 2-(3-methoxyphenyl)ethanamine
SMILES COC1=CC=CC(=C1)CCN

Specifications

Color Yellow
Boiling Point 119°C
Quantity 5 g
UN Number 2735
Formula Weight 151.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(3-Methoxyphenyl)ethylamine
RTECSNumber SH7835000
TSCA No