missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(3-Methoxyphenyl)ethylamine 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 119°C |
| Quantity | 25 g |
| UN Number | 2735 |
| Formula Weight | 151.21 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-(3-Methoxyphenyl)ethylamine |
Safety and Handling
| RTECSNumber | SH7835000 |
| TSCA | No |