missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Methoxyallylidene)malonic acid dimethyl ester, 95%, Thermo Scientific™

Chemical Identifiers

CAS 41530-32-9
Molecular Formula C9H12O5
Molecular Weight (g/mol) 200.19
InChI Key NOBNTZJTOBIOHL-GQCTYLIASA-N
Synonym 2-3-methoxyallylidene malonic acid dimethyl ester, dimethyl 2-3-methoxyallylidene malonate, 1,3-dimethyl 2-2e-3-methoxyprop-2-en-1-ylidene propanedioate, 2-3-methoxyallylidene malonic acid dimethyl ester, 96, dimethyl 2-e-3-methoxyprop-2-enylidene propanedioate, propanedioic acid, 3-methoxy-2-propenylidene-, dimethyl ester, dimethyl e-2-3-methoxyallylidene malonate, 3-methoxy-2-propenylidene malonic acid dimethyl ester, dimethyl 2-2e-3-methoxy-2-propenylidene malonate #
PubChem CID 5374172
IUPAC Name dimethyl 2-[(E)-3-methoxyprop-2-enylidene]propanedioate
SMILES COC=CC=C(C(=O)OC)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC302090025
View Documents Promotion Details
N/A 2.5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC302090100
View Documents Promotion Details
N/A 10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

41530-32-9
200.19
2-3-methoxyallylidene malonic acid dimethyl ester, dimethyl 2-3-methoxyallylidene malonate, 1,3-dimethyl 2-2e-3-methoxyprop-2-en-1-ylidene propanedioate, 2-3-methoxyallylidene malonic acid dimethyl ester, 96, dimethyl 2-e-3-methoxyprop-2-enylidene propanedioate, propanedioic acid, 3-methoxy-2-propenylidene-, dimethyl ester, dimethyl e-2-3-methoxyallylidene malonate, 3-methoxy-2-propenylidene malonic acid dimethyl ester, dimethyl 2-2e-3-methoxy-2-propenylidene malonate #
dimethyl 2-[(E)-3-methoxyprop-2-enylidene]propanedioate
C9H12O5
NOBNTZJTOBIOHL-GQCTYLIASA-N
5374172
COC=CC=C(C(=O)OC)C(=O)OC

Specifications

41530-32-9
Authentic
C9H12O5
2-3-methoxyallylidene malonic acid dimethyl ester, dimethyl 2-3-methoxyallylidene malonate, 1,3-dimethyl 2-2e-3-methoxyprop-2-en-1-ylidene propanedioate, 2-3-methoxyallylidene malonic acid dimethyl ester, 96, dimethyl 2-e-3-methoxyprop-2-enylidene propanedioate, propanedioic acid, 3-methoxy-2-propenylidene-, dimethyl ester, dimethyl e-2-3-methoxyallylidene malonate, 3-methoxy-2-propenylidene malonic acid dimethyl ester, dimethyl 2-2e-3-methoxy-2-propenylidene malonate #
COC=CC=C(C(=O)OC)C(=O)OC
200.19
200.19
2-(3-Methoxyallylidene)malonic acid dimethyl ester
120.0°C to 125.0°C (0.3 mmHg)
94% min. (GC)
1.5370 to 1.5390
NOBNTZJTOBIOHL-GQCTYLIASA-N
dimethyl 2-[(E)-3-methoxyprop-2-enylidene]propanedioate
5374172
95%
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only