missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Methoxyallylidene)malonic acid dimethyl ester, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 302090025

Quantity 2.5g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC302090025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

41530-32-9
200.19
2-3-methoxyallylidene malonic acid dimethyl ester, dimethyl 2-3-methoxyallylidene malonate, 1,3-dimethyl 2-2e-3-methoxyprop-2-en-1-ylidene propanedioate, 2-3-methoxyallylidene malonic acid dimethyl ester, 96, dimethyl 2-e-3-methoxyprop-2-enylidene propanedioate, propanedioic acid, 3-methoxy-2-propenylidene-, dimethyl ester, dimethyl e-2-3-methoxyallylidene malonate, 3-methoxy-2-propenylidene malonic acid dimethyl ester, dimethyl 2-2e-3-methoxy-2-propenylidene malonate #
dimethyl 2-[(E)-3-methoxyprop-2-enylidene]propanedioate
C9H12O5
NOBNTZJTOBIOHL-GQCTYLIASA-N
5374172
COC=CC=C(C(=O)OC)C(=O)OC

Specifications

2-(3-Methoxyallylidene)malonic acid dimethyl ester
120.0°C to 125.0°C (0.3 mmHg)
94% min. (GC)
C9H12O5
2.5g
NOBNTZJTOBIOHL-GQCTYLIASA-N
dimethyl 2-[(E)-3-methoxyprop-2-enylidene]propanedioate
5374172
95%
41530-32-9
Authentic
Glass bottle
1.5370 to 1.5390
2-3-methoxyallylidene malonic acid dimethyl ester, dimethyl 2-3-methoxyallylidene malonate, 1,3-dimethyl 2-2e-3-methoxyprop-2-en-1-ylidene propanedioate, 2-3-methoxyallylidene malonic acid dimethyl ester, 96, dimethyl 2-e-3-methoxyprop-2-enylidene propanedioate, propanedioic acid, 3-methoxy-2-propenylidene-, dimethyl ester, dimethyl e-2-3-methoxyallylidene malonate, 3-methoxy-2-propenylidene malonic acid dimethyl ester, dimethyl 2-2e-3-methoxy-2-propenylidene malonate #
COC=CC=C(C(=O)OC)C(=O)OC
200.19
200.19
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...