missing translation for 'onlineSavingsMsg'
Learn More

2,3-Heptanedione 98.0+%, TCI America™

Catalog No. H04225ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04225ML 5 mL
H042225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H04225ML Supplier TCI America Supplier No. H04225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 96-04-8
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00036550
InChI Key FJPGAMCQJNLTJC-UHFFFAOYSA-N
Synonym 2,3-heptanedione, acetyl valeryl, valerylacetyl, acetylvaleryl, acetyl pentanoyl, unii-dk55dde86p, dk55dde86p, benzil-related compound, 47, heptanedione, fema no. 2543
PubChem CID 60983
IUPAC Name heptane-2,3-dione
SMILES CCCCC(=O)C(=O)C

Specifications

Color Yellow
Boiling Point 64°C
Quantity 5 mL
UN Number 1224
Formula Weight 128.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Heptanedione
EINECSNumber (2)-0563
TSCA Yes
Recommended Storage Refrigerator