missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diphenyl-5-carboxytetrazolium Chloride 96.0+%, TCI America™

Catalog No. B0295100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0295100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0295100MG Supplier TCI America Supplier No. B0295100MG

Chemical Identifiers

CAS 2118-40-3
Molecular Formula C14H11ClN4O2
Molecular Weight (g/mol) 302.72
MDL Number MFCD00060087
InChI Key FNBVGOXTOCDSJI-UHFFFAOYSA-N
PubChem CID 131674670
IUPAC Name diphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium-5-carboxylate hydrochloride
SMILES Cl.[O-]C(=O)C1=NN(C2=CC=CC=C2)[N+](=N1)C1=CC=CC=C1

Specifications

Quantity 100 mg
Formula Weight 302.72
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Diphenyl-5-carboxytetrazolium Chloride