accessibility menu, dialog, popup

UserName

NNNNOOHCl

CAS RN 2118-40-3

IUPAC Name: diphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium-5-carboxylate hydrochloride
Synonyms: diphenyl-1,2λ⁵,3,4-tetrazol-2-ylium-5-carboxylate hydrochloride
Molecular Weight (g/mol): 302.72
Molecular Formula: C14H11ClN4O2
InChi Key: FNBVGOXTOCDSJI-UHFFFAOYSA-N
SMILES: Cl.[O-]C(=O)C1=NN(C2=CC=CC=C2)[N+](=N1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results