missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dimethylquinoxaline 98.0+%, TCI America™
$165.90 - $477.19
Chemical Identifiers
| CAS | 2379-55-7 |
|---|---|
| Molecular Formula | C10H10N2 |
| Molecular Weight (g/mol) | 158.204 |
| MDL Number | MFCD00006728 |
| InChI Key | FKHNZQFCDGOQGV-UHFFFAOYSA-N |
| PubChem CID | 16925 |
| IUPAC Name | 2,3-dimethylquinoxaline |
| SMILES | CC1=NC2=CC=CC=C2N=C1C |
Chemical Identifiers
| 2379-55-7 | |
| 158.204 | |
| FKHNZQFCDGOQGV-UHFFFAOYSA-N | |
| 2,3-dimethylquinoxaline |
| C10H10N2 | |
| MFCD00006728 | |
| 16925 | |
| CC1=NC2=CC=CC=C2N=C1C |
Specifications
| 2379-55-7 | |
| White-Yellow | |
| C10H10N2 | |
| 25 g | |
| FKHNZQFCDGOQGV-UHFFFAOYSA-N | |
| 2,3-dimethylquinoxaline | |
| 16925 | |
| ≥98.0% (T) | |
| 2,3-Dimethylquinoxaline |
| 107°C | |
| 130°C | |
| MFCD00006728 | |
| quinoxaline, 2,3-dimethyl, aurora ka-3449, quinoxaline,3-dimethyl, 2,3-dimethyl-quinoxalin, 2,3-dimethyl-quinoxaline, acmc-1cg5s, quinoxaline,2,3-dimethyl, salor-int l498777-1ea, 5-23-07-00267 beilstein handbook reference, chembl67865 | |
| CC1=NC2=CC=CC=C2N=C1C | |
| 158.204 | |
| 158.20 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : VD1960000
TSCA : Yes