accessibility menu, dialog, popup

UserName

H3CNNH3C

CAS RN 2379-55-7

IUPAC Name: 2,3-dimethylquinoxaline
Synonyms: quinoxaline, 2,3-dimethyl-
Molecular Weight (g/mol): 158.2
Molecular Formula: C10H10N2
InChi Key: FKHNZQFCDGOQGV-UHFFFAOYSA-N
SMILES: CC1=C(C)N=C2C=CC=CC2=N1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Physical Form 
  • (4)
  • (2)
Quantity 
  • (1)
  • (3)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)