Learn More
2,3-Dimethylquinoxaline, 97%
CAS: 2379-55-7 | C10H10N2 | 158.2 g/mol
$247.42 - $247.42
Chemical Identifiers
| CAS | 2379-55-7 |
|---|---|
| MDL Number | MFCD00006728 |
| InChI Key | FKHNZQFCDGOQGV-UHFFFAOYSA-N |
| PubChem CID | 16925 |
| IUPAC Name | 2,3-dimethylquinoxaline |
| SMILES | CC1=NC2=CC=CC=C2N=C1C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC116780250
|
Thermo Scientific Chemicals
116780250 |
25 g |
Each for $247.42
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2379-55-7 | |
| FKHNZQFCDGOQGV-UHFFFAOYSA-N | |
| 2,3-dimethylquinoxaline |
| MFCD00006728 | |
| 16925 | |
| CC1=NC2=CC=CC=C2N=C1C |
Specifications
| 2379-55-7 | |
| 100.0 | |
| Beige | |
| >100°C | |
| 97% | |
| C10H10N2 | |
| 25 g | |
| quinoxaline, 2,3-dimethyl, aurora ka-3449, quinoxaline,3-dimethyl, 2,3-dimethyl-quinoxalin, 2,3-dimethyl-quinoxaline, acmc-1cg5s, quinoxaline,2,3-dimethyl, salor-int l498777-1ea, 5-23-07-00267 beilstein handbook reference, chembl67865 | |
| FKHNZQFCDGOQGV-UHFFFAOYSA-N | |
| 2,3-dimethylquinoxaline | |
| 16925 | |
| 97% | |
| 2, 3-Dimethylquinoxaline, 97% |
| 96.0 | |
| 102.0°C to 107.0°C | |
| 257.0°C to 259.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD00006728 | |
| 23, 191 | |
| Solubility in water: insoluble | |
| CC1=NC2=CC=CC=C2N=C1C | |
| 158.2 | |
| 158.2 | |
| Crystalline Powder |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear prote
GHS Signal Word: Danger
EINECSNumber : 219-162-0
RTECSNumber : VD1960000
TSCA : TSCA
RUO – Research Use Only