missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylquinoxaline, 97%

CAS: 2379-55-7 | C10H10N2 | 158.204 g/mol

$58.78 - $165.41

Chemical Identifiers

CAS 2379-55-7
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.204
MDL Number MFCD00006728
InChI Key FKHNZQFCDGOQGV-UHFFFAOYSA-N
PubChem CID 16925
IUPAC Name 2,3-dimethylquinoxaline
SMILES CC1=NC2=CC=CC=C2N=C1C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2331306
View Documents
Thermo Scientific Chemicals
B2331306
5 g
Each for $58.78
Only null left
 
AAB2331314
View Documents
Thermo Scientific Chemicals
B2331314
25 g
Each for $165.41
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2379-55-7
158.204
FKHNZQFCDGOQGV-UHFFFAOYSA-N
2,3-dimethylquinoxaline
C10H10N2
MFCD00006728
16925
CC1=NC2=CC=CC=C2N=C1C

Specifications

2379-55-7
C10H10N2
5 g
quinoxaline, 2,3-dimethyl, aurora ka-3449, quinoxaline,3-dimethyl, 2,3-dimethyl-quinoxalin, 2,3-dimethyl-quinoxaline, acmc-1cg5s, quinoxaline,2,3-dimethyl, salor-int l498777-1ea, 5-23-07-00267 beilstein handbook reference, chembl67865
CC1=NC2=CC=CC=C2N=C1C
158.204
158.2
2,3-Dimethylquinoxaline
102°C to 107°C
MFCD00006728
114832
FKHNZQFCDGOQGV-UHFFFAOYSA-N
2,3-dimethylquinoxaline
16925
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 219-162-0

RTECSNumber : VD1960000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only