missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dimethylpyrazine 98.0+%, TCI America™
Supplier: TCI America D152525ML
Specifications
| 2,3-Dimethylpyrazine | |
| Yellow | |
| C6H8N2 | |
| 25 mL | |
| pyrazine, 2,3-dimethyl, 2,3-dimethyl-1,4-diazine, 2,3-dimethyl-pyrazine, pyrazine, dimethyl, 2,3-dimethyl pyrazine, ccris 2928, unii-whf7883d0v, fema no. 3271, natural, dimethylpyrazine | |
| CC1=NC=CN=C1C | |
| 108.14 | |
| 108.14 | |
| Liquid |
| 5910-89-4 | |
| 156°C | |
| MFCD00006144 | |
| 1993 | |
| OXQOBQJCDNLAPO-UHFFFAOYSA-N | |
| 2,3-dimethylpyrazine | |
| 22201 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 5910-89-4 | |
| 108.14 | |
| OXQOBQJCDNLAPO-UHFFFAOYSA-N | |
| 22201 | |
| CC1=NC=CN=C1C |
| C6H8N2 | |
| MFCD00006144 | |
| pyrazine, 2,3-dimethyl, 2,3-dimethyl-1,4-diazine, 2,3-dimethyl-pyrazine, pyrazine, dimethyl, 2,3-dimethyl pyrazine, ccris 2928, unii-whf7883d0v, fema no. 3271, natural, dimethylpyrazine | |
| 2,3-dimethylpyrazine |
Safety and Handling
EINECSNumber : (5)-0950
RTECSNumber : UQ2625000
TSCA : Yes