missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylpyrazine 98.0+%, TCI America™

Catalog No. p-7126767
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15255ML 5 mL
D152525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15255ML Supplier TCI America Supplier No. D15255ML
Only null left

Chemical Identifiers

CAS 5910-89-4
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00006144
InChI Key OXQOBQJCDNLAPO-UHFFFAOYSA-N
Synonym pyrazine, 2,3-dimethyl, 2,3-dimethyl-1,4-diazine, 2,3-dimethyl-pyrazine, pyrazine, dimethyl, 2,3-dimethyl pyrazine, ccris 2928, unii-whf7883d0v, fema no. 3271, natural, dimethylpyrazine
PubChem CID 22201
IUPAC Name 2,3-dimethylpyrazine
SMILES CC1=NC=CN=C1C

Specifications

Color Yellow
Boiling Point 156°C
Quantity 5 mL
UN Number 1993
Formula Weight 108.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Dimethylpyrazine
EINECSNumber (5)-0950
RTECSNumber UQ2625000
TSCA Yes