missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylphenylhydrazine hydrochloride, 97%

Catalog No. p-7023206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1652606 5 g
AAA1652614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1652606 Supplier Thermo Scientific Chemicals Supplier No. A1652606

CAS: 56737-75-8 | C8H13ClN2 | 172.656 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56737-75-8
Molecular Formula C8H13ClN2
Molecular Weight (g/mol) 172.656
MDL Number MFCD00013380
InChI Key UKAYNIJBIXNDDJ-UHFFFAOYSA-N
Synonym 2,3-dimethylphenylhydrazine hydrochloride, 2,3-dimethylphenyl hydrazine hydrochloride, 1-2,3-dimethylphenyl hydrazine hydrochloride, acmc-1atp8, 3-hydrazino-o-xylene hydrochloride, 2,3-dimethylphenylhydrazine, chloride, hydrazine, 2,3-dimethylphenyl-, hydrochloride 1:1
PubChem CID 2774885
IUPAC Name (2,3-dimethylphenyl)hydrazine;hydrochloride
SMILES CC1=C(C(=CC=C1)NN)C.Cl

Specifications

Melting Point ∼210°C (decomposition)
Quantity 5 g
Beilstein 6096287
Formula Weight 172.66
Percent Purity 97%
Chemical Name or Material 2,3-Dimethylphenylhydrazine hydrochloride
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
RTECSNumber MV3262500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only