missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylphenylacetic acid, 95%

Catalog No. p-7055383
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6394703 1 g
AAH6394706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6394703 Supplier Thermo Scientific Chemicals Supplier No. H6394703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 30981-98-7 | C10H12O2 | 164.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 30981-98-7
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD03844736
InChI Key AMPNBNGNPLXMOP-UHFFFAOYSA-N
Synonym 2,3-dimethylphenylacetic acid, 2-2,3-dimethylphenyl acetic acid, 2,3-dimethylbenzeneacetic acid, dimethylbenzeneacetic acid, 2,3-dimethylphenyl acetic acid, 2,3-dimethylphenylaceticacid, xylene-acetic acid, pubchem2466, acmc-1cldr, 2-2,3-dimethylphenyl aceticacid
PubChem CID 20572731
IUPAC Name 2-(2,3-dimethylphenyl)acetic acid
SMILES CC1=C(C)C(CC(O)=O)=CC=C1

Specifications

Color White
Quantity 1 g
Physical Form Powder
Formula Weight 164.2
Percent Purity 95%
Chemical Name or Material 2,3-Dimethylphenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only