missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dimethylphenol 98.0+%, TCI America™
$83.89 - $602.10
Chemical Identifiers
| CAS | 526-75-0 |
|---|---|
| Molecular Formula | C8H10O |
| Molecular Weight (g/mol) | 122.167 |
| MDL Number | MFCD00002227 |
| InChI Key | QWBBPBRQALCEIZ-UHFFFAOYSA-N |
| Synonym | 1-hydroxy-2,3-dimethylbenzene, 2,3-xylenol, dimethylphenol, xylenol, phenol, 2,3-dimethyl, o-xylenol, vic-o-xylenol, o-3-xylenol, xyellenol 100, 2,3-dimethyl phenol |
| PubChem CID | 10687 |
| IUPAC Name | 2,3-dimethylphenol |
| SMILES | CC1=C(C(=CC=C1)O)C |
Chemical Identifiers
| 526-75-0 | |
| 122.167 | |
| QWBBPBRQALCEIZ-UHFFFAOYSA-N | |
| 10687 | |
| CC1=C(C(=CC=C1)O)C |
| C8H10O | |
| MFCD00002227 | |
| 1-hydroxy-2,3-dimethylbenzene, 2,3-xylenol, dimethylphenol, xylenol, phenol, 2,3-dimethyl, o-xylenol, vic-o-xylenol, o-3-xylenol, xyellenol 100, 2,3-dimethyl phenol | |
| 2,3-dimethylphenol |
Specifications
| 526-75-0 | |
| White-Yellow | |
| C8H10O | |
| 25 g | |
| 1-hydroxy-2,3-dimethylbenzene, 2,3-xylenol, dimethylphenol, xylenol, phenol, 2,3-dimethyl, o-xylenol, vic-o-xylenol, o-3-xylenol, xyellenol 100, 2,3-dimethyl phenol | |
| CC1=C(C(=CC=C1)O)C | |
| 122.167 | |
| 122.17 | |
| Crystalline Powder |
| 75°C | |
| 218°C | |
| MFCD00002227 | |
| 2261 | |
| QWBBPBRQALCEIZ-UHFFFAOYSA-N | |
| 2,3-dimethylphenol | |
| 10687 | |
| ≥98.0% (GC) | |
| 2,3-Dimethylphenol |
Safety and Handling
EINECSNumber : (3)-0521&(4)-0057
RTECSNumber : ZE5500000
TSCA : Yes