missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylmaleic Anhydride 98.0+%, TCI America™

Catalog No. p-7127747
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
D34735G 5 g
D347325G 25 g
2 options

Catalog No. D34735G

Supplier: TCI America D34735G

Only null left

Chemical Identifiers

CAS 766-39-2
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.111
MDL Number MFCD00005523
InChI Key MFGALGYVFGDXIX-UHFFFAOYSA-N
Synonym 2,3-dimethylmaleic anhydride, dimethylmaleic anhydride, 2,5-furandione, 3,4-dimethyl, pyrocinchonic anhydride, maleic anhydride, dimethyl, dimethylmaleic acid anhydride, 2,3-dimethylmaleicanhydride, unii-6pp3n541qa, 3,4-dimethyl-furan-2,5-dione, alpha,beta-dimethylmaleic anhydride
PubChem CID 13010
IUPAC Name 3,4-dimethylfuran-2,5-dione
SMILES CC1=C(C(=O)OC1=O)C

Specifications

Melting Point 95°C
Color White-Yellow
Boiling Point 119°C
Quantity 5 g
Formula Weight 126.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dimethylmaleic Anhydride
EINECSNumber (9)-0718
RTECSNumber ON4025000
TSCA Yes