accessibility menu, dialog, popup

UserName

CAS RN 766-39-2

IUPAC Name: dimethyl-2,5-dihydrofuran-2,5-dione
Synonyms: 2,5-furandione, 3,4-dimethyl-
Molecular Weight (g/mol): 126.11
Molecular Formula: C6H6O3
InChi Key: MFGALGYVFGDXIX-UHFFFAOYSA-N
SMILES: CC1=C(C)C(=O)OC1=O
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)