missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylmaleic Anhydride 98.0+%, TCI America™

Catalog No. D34735G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34735G 5 g
D347325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34735G Supplier TCI America Supplier No. D34735G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 766-39-2
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.111
MDL Number MFCD00005523
InChI Key MFGALGYVFGDXIX-UHFFFAOYSA-N
Synonym 2,3-dimethylmaleic anhydride, dimethylmaleic anhydride, 2,5-furandione, 3,4-dimethyl, pyrocinchonic anhydride, maleic anhydride, dimethyl, dimethylmaleic acid anhydride, 2,3-dimethylmaleicanhydride, unii-6pp3n541qa, 3,4-dimethyl-furan-2,5-dione, alpha,beta-dimethylmaleic anhydride
PubChem CID 13010
IUPAC Name 3,4-dimethylfuran-2,5-dione
SMILES CC1=C(C(=O)OC1=O)C

Specifications

Melting Point 95°C
Color White-Yellow
Boiling Point 119°C
Quantity 5 g
Formula Weight 126.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dimethylmaleic Anhydride
EINECSNumber (9)-0718
RTECSNumber ON4025000
TSCA Yes