Learn More
2,3-Dimethylhydroquinone, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 305110250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 2, 3-Dimethylhydroquinone | |
| 98.5 | |
| Authentic | |
| Glass bottle | |
| MFCD00009997 | |
| 2,3-dimethylhydroquinone, o-xylene-3,6-diol, 2,3-xylohydroquinone, o-xylohydroquinone, 1,4-benzenediol, 2,3-dimethyl, unii-f0l7hg609j, 1,4-dihydroxy-2,3-dimethylbenzene, 3,6-dihydroxy-o-xylene, 2,3-dimethyl-1,4-benzenediol, 2,3-dimethyl hydroquinone | |
| BXJGUBZTZWCMEX-UHFFFAOYSA-N | |
| 2,3-dimethylbenzene-1,4-diol | |
| 69100 | |
| 97% |
| 608-43-5 | |
| 100.0 | |
| 96% min. (HPLC) | |
| C8H10O2 | |
| 25g | |
| Solubility in water: slightly soluble in water | |
| CC1=C(C)C(O)=CC=C1O | |
| 138.17 | |
| 138.17 |
Chemical Identifiers
| 608-43-5 | |
| 138.17 | |
| BXJGUBZTZWCMEX-UHFFFAOYSA-N | |
| 69100 | |
| CC1=C(C)C(O)=CC=C1O |
| C8H10O2 | |
| MFCD00009997 | |
| 2,3-dimethylhydroquinone, o-xylene-3,6-diol, 2,3-xylohydroquinone, o-xylohydroquinone, 1,4-benzenediol, 2,3-dimethyl, unii-f0l7hg609j, 1,4-dihydroxy-2,3-dimethylbenzene, 3,6-dihydroxy-o-xylene, 2,3-dimethyl-1,4-benzenediol, 2,3-dimethyl hydroquinone | |
| 2,3-dimethylbenzene-1,4-diol |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
RUO â Research Use Only