missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylfuran, 99%

CAS: 14920-89-9 | C6H8O | 96.13 g/mol

$185.36 - $185.36

Chemical Identifiers

CAS 14920-89-9
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.13
MDL Number MFCD00153893
InChI Key FJSKXQVRKZTKSI-UHFFFAOYSA-N
Synonym furan, dimethyl, dimethylfuran, dimethyl furane, dimethyl furan, 2,3-dimethyl-furan, furan, 2,3-dimethyl, 4,5-dimethylfuran, 2,3-dimethylfuran, acmc-1c70a
PubChem CID 34337
IUPAC Name 2,3-dimethylfuran
SMILES CC1=C(C)C=CO1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1570506
View Documents
Thermo Scientific Chemicals
L1570506
5 g
Each for $185.36
Only null left
 
Description

Description

2,3-Dimethylfuran may be used in the synthesis of 2,9-dioxabicyclo[3.3.1]nonane. Ireland Alcohol was synthesized in from 2,3-dimethylfuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dimethylfuran may be used in the synthesis of 2,9-dioxabicyclo[3.3.1]nonane. Ireland Alcohol was synthesized in from 2,3-dimethylfuran.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

14920-89-9
96.13
FJSKXQVRKZTKSI-UHFFFAOYSA-N
34337
CC1=C(C)C=CO1
C6H8O
MFCD00153893
furan, dimethyl, dimethylfuran, dimethyl furane, dimethyl furan, 2,3-dimethyl-furan, furan, 2,3-dimethyl, 4,5-dimethylfuran, 2,3-dimethylfuran, acmc-1c70a
2,3-dimethylfuran

Specifications

14920-89-9
42°C (115 mmHg)
C6H8O
MFCD00153893
UN1992
furan, dimethyl, dimethylfuran, dimethyl furane, dimethyl furan, 2,3-dimethyl-furan, furan, 2,3-dimethyl, 4,5-dimethylfuran, 2,3-dimethylfuran, acmc-1c70a
FJSKXQVRKZTKSI-UHFFFAOYSA-N
2,3-dimethylfuran
34337
99%
0.91
−1°C (30°F)
1.443
5 g
104572
Insoluble in water.
CC1=C(C)C=CO1
96.13
96.13
2,3-Dimethylfuran
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H301-H331
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic if inhaled.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P333+P313-P363-P370+P378q-P501c

H225-H302+H312+H332-H315-H317-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only