Learn More
2,3-Dimethylfuran, 99%
CAS: 14920-89-9 | C6H8O | 96.13 g/mol
Supplier: Thermo Scientific Chemicals L1570506
Description
2,3-Dimethylfuran may be used in the synthesis of 2,9-dioxabicyclo[3.3.1]nonane. Ireland Alcohol was synthesized in from 2,3-dimethylfuran.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,3-Dimethylfuran may be used in the synthesis of 2,9-dioxabicyclo[3.3.1]nonane. Ireland Alcohol was synthesized in from 2,3-dimethylfuran.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
| 2,3-Dimethylfuran | |
| 0.91 | |
| −1°C (30°F) | |
| 1.443 | |
| 5 g | |
| 104572 | |
| Insoluble in water. | |
| CC1=C(C)C=CO1 | |
| 96.13 | |
| 96.13 |
| 14920-89-9 | |
| 42°C (115 mmHg) | |
| C6H8O | |
| MFCD00153893 | |
| UN1992 | |
| furan, dimethyl, dimethylfuran, dimethyl furane, dimethyl furan, 2,3-dimethyl-furan, furan, 2,3-dimethyl, 4,5-dimethylfuran, 2,3-dimethylfuran, acmc-1c70a | |
| FJSKXQVRKZTKSI-UHFFFAOYSA-N | |
| 2,3-dimethylfuran | |
| 34337 | |
| 99% |
Chemical Identifiers
| 14920-89-9 | |
| 96.13 | |
| FJSKXQVRKZTKSI-UHFFFAOYSA-N | |
| 34337 | |
| CC1=C(C)C=CO1 |
| C6H8O | |
| MFCD00153893 | |
| furan, dimethyl, dimethylfuran, dimethyl furane, dimethyl furan, 2,3-dimethyl-furan, furan, 2,3-dimethyl, 4,5-dimethylfuran, 2,3-dimethylfuran, acmc-1c70a | |
| 2,3-dimethylfuran |
Safety and Handling
GHS H Statement
H225-H301-H331
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic if inhaled.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P333+P313-P363-P370+P378q-P501c
H225-H302+H312+H332-H315-H317-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only