missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylcyclohexanol (mixture of isomers) 98.0+%, TCI America™

Catalog No. D16605ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16605ML 5 mL
D166025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16605ML Supplier TCI America Supplier No. D16605ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1502-24-5
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00001444
InChI Key KMVFQKNNDPKWOX-UHFFFAOYSA-N
PubChem CID 102631
IUPAC Name 2,3-dimethylcyclohexan-1-ol
SMILES CC1CCCC(C1C)O

Specifications

Color Colorless
Boiling Point 181°C
Quantity 5 mL
Formula Weight 128.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Dimethylcyclohexanol (mixture of isomers)