accessibility menu, dialog, popup

UserName

CH3HOH3C

CAS RN 1502-24-5

IUPAC Name: 2,3-dimethylcyclohexan-1-ol
Synonyms: 2,3-dimethylcyclohexan-1-ol
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: KMVFQKNNDPKWOX-UHFFFAOYNA-N
SMILES: CC1CCCC(O)C1C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results