missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylbutane 98.0+%, TCI America™

Supplier:  TCI America D0690500ML

 View more versions of this product

Catalog No. D0690500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,3-Dimethylbutane
Colorless
500 mL
C6H14
2457
ZFFMLCVRJBZUDZ-UHFFFAOYSA-N
2,3-dimethylbutane
6589
≥98.0% (GC)
79-29-8
58°C
Clear Liquid at 20°C
MFCD00008925
diisopropyl, biisopropyl, butane, 2,3-dimethyl, 1,1,2,2-tetramethylethane, hsdb 76, unii-68isq7a432, ccris 6020, ch3 2chch ch3 2, acmc-1bmfq, dsstox_cid_5112
CC(C)C(C)C
86.178
86.18

Chemical Identifiers

79-29-8
86.178
ZFFMLCVRJBZUDZ-UHFFFAOYSA-N
6589
CC(C)C(C)C
C6H14
MFCD00008925
diisopropyl, biisopropyl, butane, 2,3-dimethyl, 1,1,2,2-tetramethylethane, hsdb 76, unii-68isq7a432, ccris 6020, ch3 2chch ch3 2, acmc-1bmfq, dsstox_cid_5112
2,3-dimethylbutane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0006

RTECSNumber : EJ9350000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions