missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylbutane 98.0+%, TCI America™

Catalog No. p-7126376
Change view
Click to view available options
Quantity:
25 mL
5 mL
500 mL
Catalog No. Quantity
D06905ML 5 mL
D069025ML 25 mL
D0690500ML 500 mL
3 options

Catalog No. D06905ML

Supplier: TCI America D06905ML

Only null left

Chemical Identifiers

CAS 79-29-8
Molecular Formula C6H14
Molecular Weight (g/mol) 86.178
MDL Number MFCD00008925
InChI Key ZFFMLCVRJBZUDZ-UHFFFAOYSA-N
Synonym diisopropyl, biisopropyl, butane, 2,3-dimethyl, 1,1,2,2-tetramethylethane, hsdb 76, unii-68isq7a432, ccris 6020, ch3 2chch ch3 2, acmc-1bmfq, dsstox_cid_5112
PubChem CID 6589
IUPAC Name 2,3-dimethylbutane
SMILES CC(C)C(C)C

Specifications

Color Colorless
Boiling Point 58°C
Quantity 5 mL
Physical Form Clear Liquid at 20°C
UN Number 2457
Formula Weight 86.18
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2,3-Dimethylbutane
EINECSNumber (2)-0006
RTECSNumber EJ9350000
TSCA Yes