missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylbutane, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019814
Change view
Click to view available options
Quantity:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019814 5 mL
111019006 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019814 Supplier Merck Emd Millipore Supplier No. 397605ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2,3-Dimethylbutane is a chemical, which can undergo photo oxidation with nitrogen oxides to give carbon monoxide and acetone as the major products.

Featured Industries: Petroleum

Specifications

Chemical Name or Material 2,3-Dimethylbutane
CAS 79-29-8
Ignition Point 788°F
Vapor Pressure 4.54 psi (25.0°C)(literature)
Vapor Density 3 (vs. air)
Density 0.662 g/mL (at 25°C)
Flash Point -29°C
Physical Form Neat
Linear Formula (CH3)2 CHCH(CH3)2
Technique Gas Chromatography
MDL Number MFCD00008925
Melting Point -129°C
Boiling Point 58°C
Synonym Diisopropyl
Type Analytical Standard
Industry Type Petrochemical Testing
Molecular Weight (g/mol) 86.18
Quantity 5 mL
Formula Weight 86.18
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less