missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylbenzonitrile 97.0+%, TCI America™

Catalog No. p-7128204
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42935G 5 g
D429325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42935G Supplier TCI America Supplier No. D42935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5724-56-1 | C9H9N | 131.18 g/mol

Chemical Identifiers

CAS 5724-56-1
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.18
MDL Number MFCD00019744
InChI Key ZEDPQIJYJCPIRM-UHFFFAOYSA-N
Synonym benzonitrile, 2,3-dimethyl, 3-cyano-o-xylene, unii-6s215f38ok, 2,3-dimethylbenzonitril german, 2,3-dimethylbenzenecarbonitrile, 2,3-dimethylbenzonitril, dimethylbenzonitrile, pubchem8125, benzonitrile, dimethyl, medetomidine impurity 11
PubChem CID 79802
IUPAC Name 2,3-dimethylbenzonitrile
SMILES CC1=CC=CC(C#N)=C1C

Specifications

Melting Point 16°C
Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
Boiling Point 62°C
Formula Weight 131.18
Percent Purity ≥97.0% (GC)
Grade Not Available
Chemical Name or Material 2,3-Dimethylbenzonitrile
RTECSNumber DI4357600
TSCA No