missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylbenzoic Acid 98.0+%, TCI America™

Catalog No. p-7127175
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22215G 5 g
D222125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22215G Supplier TCI America Supplier No. D22215G
Only null left

Chemical Identifiers

CAS 603-79-2
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002479
InChI Key RIZUCYSQUWMQLX-UHFFFAOYSA-N
Synonym hemellitic acid, benzoic acid, 2,3-dimethyl, 2,3-dimethylbenzoicacid, 2,3-dimethyl benzoic acid, 2,3-dimethyl-benzoic acid, unii-7irp8ca267, 2,3-dimethylbenzenecarboxylic acid, vic-o-xylylic acid, methyl m-toluic acid, pubchem2549
PubChem CID 11782
ChEBI CHEBI:64823
IUPAC Name 2,3-dimethylbenzoic acid
SMILES CC1=CC=CC(=C1C)C(=O)O

Specifications

Melting Point 145°C
Color White
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 150.18
Percent Purity ≥98.0% (T)
Chemical Name or Material 2,3-Dimethylbenzoic Acid
RTECSNumber DG8734000
TSCA No