missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylbenzoic acid, 98%

Catalog No. AAA1038306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1038306 5 g
AAA1038314 25 g
AAA1038322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1038306 Supplier Thermo Scientific Chemicals Supplier No. A1038306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 603-79-2 | C9H10O2 | 150.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 603-79-2
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002479
InChI Key RIZUCYSQUWMQLX-UHFFFAOYSA-N
Synonym hemellitic acid, benzoic acid, 2,3-dimethyl, 2,3-dimethylbenzoicacid, 2,3-dimethyl benzoic acid, 2,3-dimethyl-benzoic acid, unii-7irp8ca267, 2,3-dimethylbenzenecarboxylic acid, vic-o-xylylic acid, methyl m-toluic acid, pubchem2549
PubChem CID 11782
ChEBI CHEBI:64823
IUPAC Name 2,3-dimethylbenzoic acid
SMILES CC1=CC=CC(=C1C)C(=O)O

Specifications

Melting Point 143°C to 147°C
Quantity 5 g
Odor Odorless
Beilstein 971590
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material 2,3-Dimethylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-058-0
RTECSNumber DG8734000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only