missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylanthracene 98.0+%, TCI America™

Supplier:  TCI America D2979200MG

Catalog No. D2979200MG

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,3-Dimethylanthracene
252°C
C16H14
OGVRJXPGSVLDRD-UHFFFAOYSA-N
2,3-dimethylanthracene
69170
≥98.0% (GC)
613-06-9
White
200 mg
CC1=CC2=CC3=CC=CC=C3C=C2C=C1C
206.288
206.29
Crystalline Powder

Chemical Identifiers

613-06-9
206.288
69170
CC1=CC2=CC3=CC=CC=C3C=C2C=C1C
C16H14
OGVRJXPGSVLDRD-UHFFFAOYSA-N
2,3-dimethylanthracene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0640

TSCA : No

SDS
Documents

Documents

Promotions

Promotions