missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylaniline 99.0+%, TCI America™

Catalog No. D066625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D066625G 25 g
D0666500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D066625G Supplier TCI America Supplier No. D066625G

Chemical Identifiers

CAS 87-59-2
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00007732
InChI Key VVAKEQGKZNKUSU-UHFFFAOYSA-N
Synonym 2,3-xylidine, 3-amino-o-xylene, benzenamine, 2,3-dimethyl, 2,3-dimethylbenzenamine, 2,3-dimethylphenylamine, 2,3-xylylamine, ortho-xylidine, vic-o-xylidine, unii-zd450abi9x, ccris 4739
PubChem CID 6893
IUPAC Name 2,3-dimethylaniline
SMILES CC1=C(C(=CC=C1)N)C

Specifications

Quantity 25 g
Physical Form Clear Liquid at 20°C
UN Number 1711
Boiling Point 114°C
Formula Weight 121.18
Percent Purity ≥99.0% (GC,T)
Chemical Name or Material 2,3-Dimethylaniline
EINECSNumber (3)-0129
RTECSNumber ZE8750000
TSCA Yes