missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylaniline, 98%

Catalog No. p-7081099
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1091836 500 g
AAA109180E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1091836 Supplier Thermo Scientific Chemicals Supplier No. A1091836

CAS: 87-59-2 | C8H11N | 121.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87-59-2
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00007732
InChI Key VVAKEQGKZNKUSU-UHFFFAOYSA-N
Synonym 2,3-xylidine, 3-amino-o-xylene, benzenamine, 2,3-dimethyl, 2,3-dimethylbenzenamine, 2,3-dimethylphenylamine, 2,3-xylylamine, ortho-xylidine, vic-o-xylidine, unii-zd450abi9x, ccris 4739
PubChem CID 6893
IUPAC Name 2,3-dimethylaniline
SMILES CC1=C(C(=CC=C1)N)C

Specifications

Melting Point 2°C to 3°C
Density 0.992
Quantity 500 g
Odor Characteristic
Refractive Index 1.568
UN Number UN1711
Beilstein 742174
Merck Index 14,10084
Boiling Point 221°C to 222°C
Formula Weight 121.18
Percent Purity 98%
Chemical Name or Material 2,3-Dimethylaniline
Show More Show Less
Hazard Category H301+H311+H331-H373
Hazard Statement GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLIDINES, LIQUID
EINECSNumber 201-755-0
RTECSNumber ZE8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only