missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dimethyl-4-nitropyridine N-Oxide 98.0+%, TCI America™
Supplier: TCI America D29055G
Specifications
| 2,3-Dimethyl-4-nitropyridine N-Oxide | |
| 92°C | |
| MFCD00065172 | |
| 2,3-dimethyl-4-nitropyridine-n-oxide, 2,3-dimethyl-4-nitropyridine 1-oxide, 2,3-dimethyl-4-nitropyridine n-oxide, 4-nitro-2,3-lutidine-n-oxide, pyridine, 2,3-dimethyl-4-nitro-, 1-oxide, 2,3-dimethyl-4-nitropyridin-1-ium-1-olate, 4-nitro-2,3-lutidine n-oxide, 4-nitro-2,3-dimethylpyridine n-oxide, 2,3-dimethyl-4-nitro-pyridine 1-oxide, 2,3-dimethyl-4-nitro-1-oxido-pyridin-1-ium | |
| CC1=C(C)[N+]([O-])=CC=C1[N+]([O-])=O | |
| 168.15 | |
| 168.15 | |
| Crystalline Powder |
| 37699-43-7 | |
| C7H8N2O3 | |
| 5 g | |
| CFMTVTYBZMKULI-UHFFFAOYSA-N | |
| 2,3-dimethyl-4-nitropyridin-1-ium-1-olate | |
| 148223 | |
| ≥98.0% (T) |
Chemical Identifiers
| 37699-43-7 | |
| 168.15 | |
| CFMTVTYBZMKULI-UHFFFAOYSA-N | |
| 148223 | |
| CC1=C(C)[N+]([O-])=CC=C1[N+]([O-])=O |
| C7H8N2O3 | |
| MFCD00065172 | |
| 2,3-dimethyl-4-nitropyridine-n-oxide, 2,3-dimethyl-4-nitropyridine 1-oxide, 2,3-dimethyl-4-nitropyridine n-oxide, 4-nitro-2,3-lutidine-n-oxide, pyridine, 2,3-dimethyl-4-nitro-, 1-oxide, 2,3-dimethyl-4-nitropyridin-1-ium-1-olate, 4-nitro-2,3-lutidine n-oxide, 4-nitro-2,3-dimethylpyridine n-oxide, 2,3-dimethyl-4-nitro-pyridine 1-oxide, 2,3-dimethyl-4-nitro-1-oxido-pyridin-1-ium | |
| 2,3-dimethyl-4-nitropyridin-1-ium-1-olate |
Safety and Handling
RTECSNumber : UT2807000
TSCA : No